Peterson Field Guides
Supplier: Houghton Mifflin
061851743X 039535370X 0395911702 039591096X 0395935962 0395904552 9780395935439 9780395906637 9780544165625
470005-056EA 46.15 CAD
470005-056 470007-408 470007-482 470007-926 470007-934 470005-054 470007-410 470007-922 470106-214 470332-364 470332-372 470332-374
Peterson Field Guides
Educational Materials Biology Educational Materials Biology Textbooks
This exceptional guide to North American insects is a benchmark against which all other guides are measured.
  • Aligned with GLOBE protocols
The Peterson 'system' of identification uses basic patterns for comparison, and arrows pointing to certain characteristics in the illustrations indicate important marks useful for identification in the field. Each is illustrated with line drawings and photographs. Softcover, over 300 pages.
Order Now
SPECIFICATIONS
Environmentally Preferable