Somso® Twice Life-Size Heart Model
Supplier: MARCUS SOMMER
HS 5
470007-342EA 2180.55 CAD
470007-342
Somso® Twice Life-Size Heart Model
Models Human Biology Models
Coded For Easy Study And Readability
  • Large Size For Better Demonstration
  • 4 Part Model Allows For Greater Visibility Of Structures
  • 5 Year Manufacturers Warranty
Twice the life size, this excellent four-part model is sectioned so that the great vessels are depicted and the atria and ventricles are open to expose the valves. It is constructed of solid vinyl and mounted on a pedestal base.