E.I. Double Concave Glass Lenses
Supplier: Wards
PH0536J-CC PH0536I-CC PH0536A-CC PH0536D-CC PH0536H-CC PH0536L-CC PH0536B-CC PH0536C-CC
470007-086EA 16 CAD
470007-086 470007-088 470016-868 470016-914 470111-268 470111-270 470104-324 470104-326
E.I. Double Concave Glass Lenses
Educational Materials Physics Educational Materials Optics Learning Activities
Individual spherical glass lenses.
  • Double concave lenses in a variety of diameters and focal lengths
  • Made of high quality, optically worked glass
  • Highly polished surface, ground edges
This optically true lens is excellent for physics classroom demonstrations in optics and light refraction.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable