Ohaus® Spring Scales
Supplier: Ohaus
80000018 80000042 80000043 80000016 80000041 80000044 80000045 80000039 80000018
73660-006CS 12480.44 CAD
73660-006 470004-632 470004-634 470005-240 470006-148 470006-150 470015-860 470203-700 470203-702
Ohaus® Spring Scales
Balances and Scales
These spring scales are designed to stand up to heavy student use.
  • Scales withstand sudden load release
  • Individually calibrated
  • Color-coded
  • High-contrast markings
Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.
Scales are color coded, and high-contrast markings make readings easy.