Spectrum Tubes
Supplier: Wards
PH1196B-5 PH1196B-3 PH1196B-7 PH1196B-2 PH1196B-4 PH1196B-1 PH1196B PH1196B-AIR PH1196B-10 PH1196B-6 PH1196B-11 PH1196B-8 STB-MX 3376-16 3376-09 PH1196B9
470005-836EA 79.1 CAD
470005-836 470014-472 470014-474 470014-478 470014-480 470014-482 470014-484 470014-486 470013-006 470014-470 470014-476 470014-702 470061-706 470163-922 470164-816 470230-432
Spectrum Tubes
Educational Materials Physics Educational Materials Optics Learning Activities
Spectrum Tubes demonstrate the wave properties of different gases.
  • Ideal for Wavelength Determination
  • For Introducing Students to Better-Known Gases
  • Glass Tube Length: 10"
  • Capillary Portion Length: 4"
  • Tubes Work in a Variety of Spectroscopic Applications
Ideal for wavelength determination and introducing students to the spectra of better-known gases, these glass tubes are 10" long, and the capillary portion is about 4" long. The spectrum tube assemblies can be used in a variety of spectroscopic applications.
When they are observed through aMore Product Information