Siren Disk
Supplier: Science First
ER-6A
470005-720EA 1315.05 CAD
470005-720
Siren Disk
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Explore the relationship between acoustical tones and the physical circumstances that give rise to them
  • Perforated rotating disk
  • Made under computer control
  • 18 x 13 x 4.5 cm
  • 23 cm H
Can be used to investigate the resonance of tubes and containers and to explore the evolution of musical scales. Comes with a sounding tube and mandrel. We recommend mounting it on a standard lab rotator such as our Universal Rotator (shown here). Uniformly spaced holes are 8, 10, 12, 16, 24, 27, 30, 32, 36, 40, 45 and 48 holes per revolution.
Order Now
SPECIFICATIONS
Environmentally Preferable