Sliding Microtome
Supplier: Uchida Yoko
162-3012 916-2304
470005-082EA 551.4 CAD
470005-082 470005-084
Sliding Microtome
Microtomes and Microtome Assemblies Microtomes
Hands-on tool for scientific studies
  • Built in safety features
  • Adjustable section thickness
Cut sections to 40-50 microns safely and easily with this microtome, which features a clear plastic safety cover, fixed blade and rugged plastic construction for hands-on student use.
Simply secure the specimen in the holder, advance using the feed screw, then draw it over the blade. The feed screw moves the specimen only 0.5 mm per complete rotation, and can be advanced as little as 10 microns. For very thin sections, the specimen can be wrapped in an artificialMore Product Information