AC/DC Generator Demo Set
Supplier: United Scientific Supplies
ACDCSET
470004-798EA 155.25 CAD
470004-798
AC/DC Generator Demo Set
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Visually impacting way to demonstrate an AC and DC generator.
  • Activity guide included
  • Clear base for easy demonstration of connections
This unique hand generator is an easy yet effective way to teach how AC and DC electricity is produced. Banana plugs can be inserted into the different connectors to change from AC to DC electricity. Simply turn the hand wheel and the included bulb will light up. The generator is a permanent-magnet rotating armature generator. The dimensions of the base are 225mm L x 145mm W x 30mm D.
Order Now
SPECIFICATIONS
Environmentally Preferable