Moment of Inertia Demonstrator
Supplier: Avantor
470003-706EA 93.05 CAD
470003-706
Moment of Inertia Demonstrator
Educational Classroom Kits and Activities
This machine, designed by teacher Bruce Yeany, offers a hands-on demonstration of the relationship between the moment of inertia and the distribution of mass.
  • Economical
  • For Grades Eight through Ten
  • Ideal for Demonstrating Inertia
  • Designed by Teacher Bruce Yeany
Hold the demonstrator in one hand, slide the attached blocks in either direction from the center of the meter stick, and rotate. By sliding the blocks toward the ends of the meter stick, students observe that it takes a certain amount of effort to rotate the demonstrator.
Now slide theMore Product Information