Microscope Slide, Double Shallow Depression
Supplier: PRECISION MOULDED POLYMERS LIMITED
1350611040004
470003-480PK 47.4 CAD
470003-480
Microscope Slide, Double Shallow Depression
Slides Microscope Slides
Use this slide to examine liquids more easily
  • Quality slide making supplies
  • Examine liquids and water borne organisms
Excellent for holding liquids and cultures for microscopic examination. The depressions are approximately 13 mm in diameter and are 0.5-0.8 mm deep.