Rheostat
Supplier: Wards
PH1155A PH1155H PH1155J PH1155F
470003-094EA 231.35 CAD
470003-094 470003-096 470003-098 470152-306
Rheostat
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This newly designed rheostat features a protective cover.
  • Oxidized Resistance Wire
  • Safety-Socket Terminals
  • Resistance: ±10%
  • Protective Cover
It's wound with a heavily oxidized resistance wire on a tube and fitted with 4 mm safety-socket terminals.
Its resistance is ±10% compared to nominal value.
Order Now
SPECIFICATIONS
Environmentally Preferable