Wind Generator
Supplier: Wards
WLS1755-57
470003-064EA 68.45 CAD
470003-064
Wind Generator
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
A small scale demonstration of renewable energy, this wind generator can be used with a voltmeter or datalogger to measure and monitor output.
  • Size: 15.2×10.2×15.2 cm
It can also be used to illuminate an LED and illustrate the conversion of energy. Its low-inertia electric motor functions as a generator and is driven by a plastic impeller.
A cold air blower can be used to turn the impeller resulting in a low voltage DC output at the two 4 mm sockets.