Formaldehyde-Free Bullfrogs
Supplier: Ward's Science
136.0801.10 136.0803.10 136.0809.10
470000-612PK 429.7 CAD
470000-612 470000-614 470000-724
Formaldehyde-Free Bullfrogs
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free bullfrogs. Available in pails or vacuum packs.
  • Amphibian anatomy
  • General biology
To better serve the needs of all teaching environments and preferences, WARD’S offers this line as an alternative to our existing line of high-quality, safe preserved specimens. Absolutely no formaldehyde whatsoever is used in the fixing and preserving process for these specimens. Instead, our new solution used for preparation binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD’SMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable