Formaldehyde-Free Grassfrogs
Supplier: Ward's Science
136.0899.50 136.0901.50 136.0903.50 136.0909.100 136.0911.100 136.0913.100 136.0900.10 136.0904.10 136.0906.10 136.0912.10
470000-522PK 620.85 CAD
470000-522 470000-524 470000-526 470000-528 470000-530 470000-532 470001-676 470001-680 470001-682 470001-686
Formaldehyde-Free Grassfrogs
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free grassfrogs. Available in pails or vacuum packs.
  • Amphibian anatomy
  • General biology
To better serve the needs of all teaching environments and preferences, WARD’S offers this line as an alternative to our existing line of high-quality, safe preserved specimens.
Absolutely no formaldehyde whatsoever is used in the fixing and preserving process for these specimens.
Instead, our new solution used for preparation binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD’SMore Product Information