Formaldehyde-Free Fetal Pigs
Supplier: Ward's Science
140.1245.10 140.1246.10 140.1247.10 140.1242.10 140.1244.10 140.1230.1 140.1232.1 140.1233.1 140.1234.1 140.1235.1
470000-434PK 478.65 CAD
470000-434 470000-438 470000-442 470000-458 470000-460 470001-716 470001-718 470001-720 470001-724 470001-728 470228-332 470228-334
Formaldehyde-Free Fetal Pigs
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free preserved fetal pigs.
  • Mammalian anatomy
  • High quality preservation
To better serve the needs of all teaching environments and preferences, WARD’S offers this line as an alternative to our existing line of high-quality, safe preserved specimens. Absolutely no formaldehyde whatsoever is used in the fixing and preserving process for these specimens. Instead, our new solution used for preparation binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD'S offers the largest selectionMore Product Information