Protector® Acid Storage Cabinets, Labconco®
Supplier: Labconco
9901600 9905300 9901400 9901500 9905200 9901300 9905000 9901200 9901100 9901000 9905100
46610-408EA 3713 CAD
46610-408 46610-410 46610-412 46610-424 46610-426 46610-428 46610-430 46610-456 46610-464 46610-472 46610-480
Protector® Acid Storage Cabinets, Labconco®
Furniture Storage Cabinets
Safely store and vent acids and other highly corrosive liquids.
  • Cabinets are constructed of durable white epoxy-coated steel with corrosion-resistant polyethylene interior liner
  • Equipped with leveling legs and rear vent connection cutouts for venting the cabinet into a laboratory hood
Include one 20.3 cm (8") filler panel to increase cabinet’s overall depth from 55.9 cm (22") to 76.2 cm (30"), for installation flush to wall.
Accessories information: Accessory interior shelf kits include powder-coated steel shelf, acid-resistant PVC tray andMore Product Information