500 Series Safety Goggles, Encon® Safety Products
Supplier: Encon Safety Products
05058204
46500-050CS 196.13 CAD
46500-050 46500-054
500 Series Safety Goggles, Encon® Safety Products
Goggles
Goggle features black neoprene strap, indirect venting, SV2™ side vent.
  • Distortion-free optics
  • Impact resistant
  • Replaceable lens
  • Four base curve lens
  • Flexible fitting
  • 99.9% UVA and UVB protection to 385 nm
These goggles have high tech and a light weight body design with superior chemical splash protection.
Certifications: NSI Z87.1+, Conformity to ANSI Z87.1-2010 has been assessed per ANSI/ISEA 125-2014 Level 3.