MBP 101 to 1000 µl Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
111-11 111-R100S 111-11 111-R 111-R100N 111-R100NS H-111-R100NS 111-RN 111-N H-111-R100
46300-442CS 1375.39 CAD
46300-442 46300-446 89404-804 89404-808 89404-810 89404-812 490008-564 490014-526 490016-322 490016-324
MBP 101 to 1000 µl Pipette Tips, Thermo Scientific
Pipette Tips
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product.
  • Metal-free Tips: All materials used in the manufacture of these tips are free of trace metals
  • A trace metal certificate is available upon request
  • A mold release agent is NOT used in the manufacturing of these pipette tips
Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non-More Product Information