Anticon 100® Standard Weight Series Cleanroom Wipers, Contec®
Supplier: Contec
495352-803 495352-805
45800-016CS 809.35 CAD
45800-016 45800-018 45800-020
Anticon 100® Standard Weight Series Cleanroom Wipers, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Anticon 100® StandardWeight™ wipes are made from 100% continuous filament polyester with a double-knit, no-run interlock construction. The clean cut edges create a very soft wipe, where the whole of the wipe fabric can be used with no risk of scratching.
  • Patented textile technology creates a polyester wipe with high sorbency
  • Lowest fibers and particles available for a cut edge wipe reduces risk of contamination
  • High sorptive capacity decreases the number of wipes required for standard use
  • Excellent choice for wiping and absorbing spills in critical
More Product Information