MicroSeal™ CSLB™ Sleeve Seal Tapes, Micronova
Supplier: Micronova
CSLB-3WH CSLB-3WHIR CSLB-12WH CSLB-1WHIR CSLB-1WH
43300-942CS 657.39 CAD
43300-942 68400-422 68400-432 68400-434 68400-436
MicroSeal™ CSLB™ Sleeve Seal Tapes, Micronova
Tapes
These cleanroom tapes are made of PE with pressure-sensitive acrylic adhesive.
  • Pre-perforated
  • Perforations at 7,6 cm (3”) intervals allow an easy tear at optimum lengths
  • Low-density, low-outgassing polyethylene film will remove cleanly, leaving no adhesive residue
  • Can be written on with fibre tip and permanent markers
  • Wound on a 3" plastic core with no liner, meaning less packaging and waste to discard
  • Available from 0.5 to 3" widths; smaller widths are ideal for rodac plate sealing during environmental monitoring
  • Suitable for food processing and
More Product Information