J.T.Baker® Plate, Dilution Strips SW
Supplier: Ritter, part of Avantor
43001-00311
43001-0031CS 629.17 CAD
43001-0031
J.T.Baker® Plate, Dilution Strips SW
Microplates Deep Well Microplates
Suitable for use on Dynex DSX and DS2 instruments. Deep well reservoir with 8 wells.
  • Strip of eight wells
  • Made of PP