VWR® Irradiated Non-Skid Shoe Covers
Supplier: VWR International
414004-475 414004-476 414004-477 414004-478 414004-479
414004-475CS 160.44 CAD
414004-475 414004-476 414004-477 414004-478 414004-479
VWR® Irradiated Non-Skid Shoe Covers
Disposable Garments
Irradiated non-skid shoe covers have been engineered to optimize skid resistance, barrier protection, cleanliness, and comfort.
  • Non-skid soles provide maximum traction on wet and smooth surfaces
  • Superior coefficient of friction
  • Constructed of impervious material
  • Clean, low-linting fabric
Shoe covers are gamma irradiated to a Sterility Assurance Level (SAL) of 10-4 to address the concern of bio-burden contamination. Manufactured in an ISO Class 8 (FED-STD-209E Class 100,000/M6.5) cleanroom.
Abrasion resistance: 1000 (sole)
Certifications: ShoeMore Product Information