VWR® Maximum Frocks
Supplier: VWR International
414004-394 414004-395 414004-396 414004-397 414004-398 414004-399 414004-695 414004-696 414004-697 414004-698 414004-699 414004-700 414004-701 414004-702
414004-394CS 430.37 CAD
414004-394 414004-395 414004-396 414004-397 414004-398 414004-399 414004-695 414004-696 414004-697 414004-698 414004-699 414004-700 414004-701 414004-702
VWR® Maximum Frocks
Disposable Garments
These are the cleanest and most durable frocks available within the VWR® Protection line.
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding
  • Light chemical-splash resistance
  • Breathable to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
Manufactured from a specially formulated breathable microporous fabric, these garments are distinguished by their exceptional comfort benefits and superior barrier protection capabilities
Garments feature raglan sleeves, and elastic cuffs. Available with snaps or zipperMore Product Information