VWR® High and Low Form Porcelain Crucibles
Supplier: VWR International
414004-231 414004-232 414004-233 89037-988 89037-990 89037-992 89037-994 89037-996 89037-998 89038-000 89038-002 89038-022 89038-024 89038-026 89038-028 89038-030 89038-034 89040-208
414004-231CS 702.33 CAD
414004-231 414004-232 414004-233 89037-988 89037-990 89037-992 89037-994 89037-996 89037-998 89038-000 89038-002 89038-022 89038-024 89038-026 89038-028 89038-030 89038-034 89040-208
VWR® High and Low Form Porcelain Crucibles
Crucibles
Chemical-porcelain crucibles are glazed inside and out, except the outside bottom and rim.
  • Glazed interior and exterior
  • Crucible covers are completely glazed, except for the rim
  • For use in temperatures up to 1150 °C (2102 °F)
Order Now
SPECIFICATIONS
Environmentally Preferable