VWR® Laboratory Bottles, LDPE
Supplier: VWR International
414004-169 414004-170 414004-171 414004-172 414004-173 414004-174 414004-175 89221-598 89221-600 89221-606 89221-608 89221-610 89221-646 89221-648 89221-650 89221-652 89221-654 89221-656
414004-169CS 127.56 CAD
414004-169 414004-170 414004-171 414004-172 414004-173 414004-174 414004-175 89221-598 89221-600 89221-606 89221-608 89221-610 89221-646 89221-648 89221-650 89221-652 89221-654 89221-656
VWR® Laboratory Bottles, LDPE
Bottles Plastic Bottles
Translucent, flexible bottles are for general laboratory use, and are especially good for shipping liquids and for trace metal analysis.
  • Leakproof
  • Translucent siding
Can be stored in freezer to −100 °C (−148 °F). Offer better visibility of contents than high-density polyethylene bottles, and have excellent chemical resistance to most acids, bases, and alcohols.
Delivery information: Polypropylene screw closures included.
Order Now
SPECIFICATIONS
Environmentally Preferable