VWR® Laboratory Bottles, HDPE
Supplier: VWR International
414004-151 414004-152 414004-153 414004-154 414004-155 414004-156 414004-157 414004-158 414004-159 89221-580 89221-582 89221-584 89221-586 89221-588 89221-590 89221-592 89221-594 89221-596 89221-658 89221-660 89221-662 89221-664 89221-666 89221-668
414004-151CS 129.17 CAD
414004-151 414004-152 414004-153 414004-154 414004-155 414004-156 414004-157 414004-158 414004-159 89221-580 89221-582 89221-584 89221-586 89221-588 89221-590 89221-592 89221-594 89221-596 89221-658 89221-660 89221-662 89221-664 89221-666 89221-668
VWR® Laboratory Bottles, HDPE
Bottles Plastic Bottles
Translucent, rigid bottles are for general laboratory use and for storing, shipping, and packing liquids.
  • Durable for long-term use
  • High-density polyethylene construction
  • Translucent siding
Can be stored in freezer to −100 °C (−148 °F). Excellent chemical resistance to most acids, bases, and alcohols. Can be used with most corrosives.
Delivery information: Polypropylene screw closure included.
Order Now
SPECIFICATIONS
Environmentally Preferable