VWR® Disposable Transfer Pipettes, Graduated
Supplier: VWR International
16001-172 16001-176 16001-180 16001-182 16001-188 414004-013 414004-014 414004-015 414004-016 414004-017 414004-024 414004-025 414004-034 414004-035 414004-040 414004-043 414004-044 414004-045
16001-172CS 770.35 CAD
16001-172 16001-176 16001-180 16001-182 16001-188 414004-013 414004-014 414004-015 414004-016 414004-017 414004-024 414004-025 414004-034 414004-035 414004-040 414004-043 414004-044 414004-045
VWR® Disposable Transfer Pipettes, Graduated
Pipettes Pasteur/Transfer Pipettes
VWR® Disposable Transfer Pipettes are ideal for quick and safe fluid transfer.
  • Will not shatter or break
  • Can be used in liquid nitrogen
  • Nontoxic and inert
  • No bulb to insert or remove
  • Uniform drop size
These unbreakable all-in-one graduated transfer pipettes eliminate the hazard of broken glass and exposure to infectious materials. Molded from see-through low density polyethylene, pipettes are inert to biological fluids and most acids, and can be sealed and refrigerated. The low-affinity surface reduces the loss of cells and valuable proteins due toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable