Accessories for Sonicator® Ultrasonic Cell Disruptors, Qsonica
Supplier: QSonica, LLC.
448
40000-716EA 759.44 CAD
40000-716
Accessories for Sonicator® Ultrasonic Cell Disruptors, Qsonica
Homogenizers Ultrasonic Cell Disruptors
A variety of horns are available to adapt Sonicator cell disruptors (see 97007-386 series) for differing sample size and intensity requirements. An increase in horn diameter decreases intensity and amplitude.
Horn extenders, also available in solid or tapped varieties, can reach into long neck vessels. Half-wave extenders add 12.7cm (5") to the horn’s length; full-wave extenders add 25.4 cm (10") to the horn’s length.
Cup horns offer high throughput, indirect sonication while eliminating cross contamination, aerosolizing, and foaming. They allow samples to be processed in sealed tubesMore Product Information