Foaming Characteristics Apparatus, Koehler
Supplier: Koehler
K43002 K43003 K43041 K43049 K43092 K43093
37000-254EA 51830.89 CAD
37000-254 37000-256 37000-262 37000-264 37000-266 37000-268
Foaming Characteristics Apparatus, Koehler
Petroleum Testing Apparatus
Lubricating oils are aerated at specified temperatures to generate a foam, which is measured at different intervals
Dual-twin and automatic time sequence models are suitable for standard foaming characteristics tests: two tests each at 24°C (75°F) and 94°C (200°F). Air flow rate is 94mL/min. Cold bath has built-in coils for circulating exit air from the high-temperature cylinder and a separate coil for circulating cooling water or refrigerant when ambient temperature exceeds the test temperature. Meet specifications: ASTM D892, IP 146, DIN 51566, FTM 791-3211, and NF T 60-129.