UVP EL Series Ultraviolet Lamps, Analytik Jena
Supplier: Analytik Jena US
95-0200-02 95-0201-02 95-0248-02 95-0249-02 95-0250-02 95-0251-02 95-0252-02 95-0198-02 95-0199-02 95-0199-01 95-0198-01 95-0248-01 95-0250-01 95-0249-01 95-0251-01 95-0264-01 95-0264-02 95-0266-01 95-0266-02 95-0267-01 95-0267-02 95-0269-02 95-0269-01 95-0271-01 95-0271-02 95-0272-01 95-0272-02 95-0273-01 95-0273-02 95-0274-01 95-0274-02 95-0277-01 95-0277-02 95-0275-01 95-0275-02 95-0276-01 95-0276-02 95-0278-01 95-0278-02 95-0279-01 95-0279-02 95-0201-01 95-0252-01
21474-566EA 1256.64 CAD
21474-566 21474-570 21474-572 21474-574 21474-576 21474-578 21474-580 21474-640 21474-642 36553-122 36553-124 36553-126 36553-128 36553-130 36553-132 36553-136 63220-240 63220-242 63220-244 63220-246 63220-248 63220-250 63220-252 63220-254 63220-256 63220-258 63220-260 63220-262 63220-264 63220-266 63220-268 63220-270 63220-272 63220-274 63220-276 63220-278 63220-280 63220-282 63220-284 63220-286 63220-288 63220-290 36553-134 36553-138
UVP EL Series Ultraviolet Lamps, Analytik Jena
Lights UV Lights, Lamps and Bulbs
The EL Series UV Lamps from Analytik Jena provide models with longwave, shortwave, midrange UV and/or white light in four, six or eight watts intensities. Some models feature multiple UV wavelengths in the same lamp.
  • Lamps are manufactured with a durable, extruded metal housing and scratch-resistant powder paint
  • A selector switch located on the lamp back makes it easy to change from one light source to the other
The duo models house one UV tube and one white light tube -- the advantage of white light or UV illumination in one unit.
Use white lightMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable