Microplate Incubator, Boekel Scientific
Supplier: Boekel
260700
35915-118EA 1096.66 CAD
35915-118
Microplate Incubator, Boekel Scientific
Incubators Microplate Incubators
Ideal for small volume incubations at or near 37°C (98.6°F)
Versatile and economical. Holds up to 14 microtiter plates. Cost-effective way to incubate petri dishes or test tubes. Although pre-set at 37°C (98.6°F), it can be adjusted from 32 to 42°C (89.6 to 107.6°F). Plexiglas® door for easy viewing of contents. Unobstructed interior allows excellent space utilization. Uniform heat is provided by a hidden element located in the back of the unit. Includes pilot lamp indicator, and unbreakable, flat plastic Digitemp® thermometer, which is attached to the chamber floor. Durable aluminumMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable