Microplate Replicators, Boekel Scientific
Supplier: Boekel
140500 140501
35915-116EA 1248.58 CAD
35915-116 70380-018
Microplate Replicators, Boekel Scientific
Microplate and Petri Dish Replicators
The microplate replicator is a convenient, time-saving device for transferring small volumes of inoculum from microplates to other microplates or solid supports.
  • Pins made of stainless steel for durability, autoclaving, and flame sterilisation
  • Economical alternative to robotic systems
  • 48- and 96-pin models include an alcohol reservoir and cleaning pads for debris removal
  • Template model available for 384-pin replicator
Applications include replication of YACs and cosmid libraries, inoculation of filters for colony hybridisation, antibiotic sensitivityMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable