VWR® Specific Gravity ASTM Hydrometers, Traceable to NIST
Supplier: VWR International
35123-302 35123-324 35123-346 35123-368 35123-380 35123-426 35123-460 35124-586 34676-180 34676-420 34676-543 35123-404 35124-600 35126-013 35126-049 35126-060 35126-082 35126-140
35123-302EA 12.24 CAD
35123-302 35123-324 35123-346 35123-368 35123-380 35123-426 35123-460 35124-586 34676-180 34676-420 34676-543 35123-404 35124-600 35126-013 35126-049 35126-060 35126-082 35126-140
VWR® Specific Gravity ASTM Hydrometers, Traceable to NIST
Hydrometers
Available in a variety of models measuring specific gravity of heavy and light liquids in various ranges from 0.650 to 2.000 in accordance with the American Society of Testing Materials (ASTM) E100 standards.
  • Conform to the accuracy requirements of ASTM hydrometers
  • Commonly used in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F/60°F
  • Hand inspected by our U.S. quality team
Packaged with a multi-language (English,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable