VWR® Specific Gravity/Relative Density (g/cm³) Precision Hydrometers, Traceable to NIST
Supplier: VWR International
34627-027 34627-050 34627-173 34627-174 34627-231 34627-253 34627-297 34627-311 34627-355 34627-388 34627-446 34627-479 34627-628 34627-082 34627-118 34627-130 34627-413 34627-504 34627-537 34627-560 34627-650
34627-027EA 116.94 CAD
34627-027 34627-050 34627-173 34627-174 34627-231 34627-253 34627-297 34627-311 34627-355 34627-388 34627-446 34627-479 34627-628 34627-082 34627-118 34627-130 34627-413 34627-504 34627-537 34627-560 34627-650
VWR® Specific Gravity/Relative Density (g/cm³) Precision Hydrometers, Traceable to NIST
Hydrometers
Available in a wide range of models measuring specific gravity/relative density (g/cmᶟ) of a variety of liquids in various ranges from 0.600 to 1.920, with 0.0005 divisions.
  • 325 mm (12.8”) long
  • Lead free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F (15°C)
  • Hand inspected by our U.S. quality team
Specific gravity hydrometers are the most widely used hydrometer, common in industries such as food and beverage, chemical, pharmacology, transportation, refrigeration and more.
Packaging: Packaged with a multi-language (More Product Information