Bromothymol blue solution

Supplier: LA MOTTE CHEMICAL PRODUCTS CO

Synonyms: 3′,3″-Dibromothymolsulphonphthalein, 3′,3″-Dibromothymolsulfonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

2210-L
34186-431EA 72.66 CAD
34186-431
Bromothymol blue solution
Bromothymol blue
Formula: C₂₇H₂₈Br₂O₅S
MW: 624.39 g/mol
MDL Number: MFCD00005872
CAS Number: 76-59-5
Merck Index: 13,01430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR