Hydrion pH Buffers, Micro Essential Lab
Supplier: MICRO ESSENTIAL LABORATORY I
2801000 2801100 2801200 280200 280400 280500 280600 280640 280686 280700 2807400 280800 280900 CC-725 280-3
34175-077PK 30.29 CAD
34175-077 34175-079 34175-081 34175-083 34175-087 34175-089 34175-091 34175-093 34175-095 34175-097 34175-099 60775-984 60775-986 60785-007 62000-406
Hydrion pH Buffers, Micro Essential Lab
Buffers pH Calibration Buffers
Chemvelopes are made of a triple protective foil consisting of aluminum, polyethylene and paper which keeps the Hydrion Buffers dry and free-flowing.
Hydrion buffer chemvelopes produce 500 ml of pH buffer standard solution when dissolved in distilled water.
Packaging: Each carton contains five chemvelopes.
Order Now
SPECIFICATIONS
Environmentally Preferable