IKA® Works Reciprocating Shakers, 230 V, HS 260
Supplier: IKA Works
3066600 3066700
33998-366EA 7383.32 CAD
33998-366 33998-370
IKA® Works Reciprocating Shakers, 230 V, HS 260
Shakers Reciprocating Shakers
These compact, flat shakers feature electronic adjustment of the speed and timer.
  • 230 V model
The basic model has a timer with a range of 5 to 50 minutes. The control model features a timer with a range of 0 to 9 hours, 59 minutes, as well as a digital display to allow users to read the speed, timer function, and operating modes at a glance. Control model also features integrated end point positioning for automated robot-controlled sampling. All functions on the control model can be operated and documented with labworldsoft® software using the RS-232More Product Information