Lab Clamp, Heavy Duty, Glas-Col®, LLC
Supplier: Glas-Col
100DLC300
33796-002EA 370.83 CAD
33796-002
Lab Clamp, Heavy Duty, Glas-Col®, LLC
Clamps General Purpose Clamps
Right angle clamp with adjustment knobs.
  • Sturdy construction
  • Easily attached
  • Easy adjustment
This right angle lab clamp is ideal for heavy duty use on 3/4" rods.
Delivery information: 7-10 days after order.