Heating Mantles, Series M, Glas-Col®
Supplier: Glas-Col
100CM120 100CM98
33787-620EA 5520.06 CAD
33787-620 33787-622
Heating Mantles, Series M, Glas-Col®
Heaters Heating Mantles
Compact aluminum housed heating mantle.
  • Fabric interior
  • Durable aluminum outer shell
  • Space saving compact design
Our Series "M" mantle is the most compact mantle with an aluminum shell. The resilient glass fabric interior supports the flask and protects it from thermal strain. Only the bottom of flask is covered so the contents are fully visible during process.
Delivery information: 7-10 days after order.
Order Now
SPECIFICATIONS
Electrical 115V
Environmentally Preferable