Cole-Parmer® SHM-200 Series Multiposition Extraction Heater/Stirrers, Antylia Scientific
Supplier: Antyila Scientific
03076-15 03076-36 03076-39 03076-42 03076-45 03076-12 03076-18 03076-21
33749-202EA 5019.8 CAD
33749-202 33749-300 33749-326 33749-359 33749-382 76045-866 76045-868 76045-870
Cole-Parmer® SHM-200 Series Multiposition Extraction Heater/Stirrers, Antylia Scientific
Heaters Heating Mantles
Allows liquids with different boiling points to be heated and/or stirred next to each other.
  • Stir or heat solutions simultaneously in each position
  • Accommodate flask capacities from 100 to 1000 ml
  • Unique vented airflow ensures the case remains 'cool-to-the-touch'
  • Robust construction permits continuous operation of the mantles as required by routine extraction tests
Regulating the heat on these multiposition extraction mantles is simple using the built-in solid-state electronic controls. The units accept round-bottom flasks ranging from 100 to 1000 ml.More Product Information