Durometers, Hardness Tester
Supplier: Pacific Transducer
306L 307L
33674-000EA 3234.92 CAD
33674-000 33676-005
Durometers, Hardness Tester
Durometers
ASTM Type A durometer measures hardness of rubber, elastomers, and other rubber-like materials including neoprene, polyurethane, silicone, vinyl, and butyl
ASTM Type D durometer is designed to measure hardness of hard rubber, plastics, Formica, Plexiglas®, soft metals, epoxies, nylon, polyurethane, and other similar materials.
Durometers meet or exceed current ASTM D2240 specifications. Accuracy of ±1 point over the entire scale. Durometers feature a maximum hold hand which stops at the peak reading. Carrying case, check block, and instructions are included.
Also measures softMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable