PYREX® Separatory Funnel, Corning
Supplier: Corning
6413-125FO
33501-410CS 1536.55 CAD
33501-410
PYREX® Separatory Funnel, Corning
Funnels Separating Funnels
These replacement 125mL PYREX® glass separatory funnels have inner and outer 19/22 Standard Taper joints
They require Polytetrafluorethylene (PTFE) No. 2 Product Standard stopcock plugs which fit into the micro-finish barrel. The plug also reduces freezing problems and grease contamination. Replacement PTFE stopcock plugs are sold seperately.