KoolTemp® Insulated Shippers, EPS Foam Shipping Coolers, Cold Chain Technologies
Supplier: Cold Chain Technologies
TL1189-SU-V TL2000-M-V TL2000-SU-V TL2300-SU-V TL2800-SU-V TL644-M-V TL867-SU-V TL645-SU-V TL1187-SU-V TL12107-SU-V TL12109-SU-V TL1500-SU-V TL3800-SU-V TL888-SU-V TL8612-M-V TL8612-SU-V TL864-SU-V TL869-M-V TL869-SU-V
33500-376PK 741.38 CAD
33500-376 33500-378 33500-380 33500-384 33500-388 33500-390 33500-404 33500-406 80081-202 80081-204 80081-206 80081-208 80081-210 80081-212 80081-214 80081-216 33500-394 33500-396 33500-400 33500-410 33500-412
KoolTemp® Insulated Shippers, EPS Foam Shipping Coolers, Cold Chain Technologies
Mailers and Shippers
Lightweight and reusable insulated expanded polystyrene (Styrofoam) shippers provide excellent thermal protection for perishables and samples.
  • Reusable
  • Excellent thermal protection
Channel construction and stand-off pads keep product away from condensation and provide more uniform temperature control throughout the cooler when used with dry ice or Koolit® refrigerants. State-of-the-art lid interlock design and secure toungue-and-groove friction fit minimize thermal loss. Rounded corners resist friction damage.
Delivery information: Shippers areMore Product Information