VWR® Manometer Pressure/Vacuum Gauges
Supplier: VWR International
33500-082 33500-086 33500-088 33500-084
33500-082EA 714.11 CAD
33500-082 33500-086 33500-088 33500-084
VWR® Manometer Pressure/Vacuum Gauges
Manometers
Differential feature registers the pressure or vacuum difference between two different sources, and memory feature recalls highest and lowest readings over any time period.
  • Traceable®
  • Measures in 11 units
A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Manometers display differential pressure or vacuum readings in 11 different units: psi, mbar, bar, cm/H2O, in./H2O, ft./H2O, mm /Hg,More Product Information