Nalgene® Graduates, Double Scale Pharmaceutical, Thermo Scientific
Supplier: Thermo Fisher Scientific
3673-0004 3673-0008 3673-0016 3673-0032
33034-100CS 669.15 CAD
33034-100 33034-121 33034-143 33034-165
Nalgene® Graduates, Double Scale Pharmaceutical, Thermo Scientific
Cylinders Measuring Cylinders
Molded of polymethylpentene
The 125, 250, 500 and 1000mL graduates conform to the volume and accuracy requirements of ASTM E1094. The 30mL graduate is also graduated in drams.
Transparent, shatterproof. Graduates feature remarkable heat tolerance to 175°C and excellent chemical resistance. Graduated in ounces and milliliters. Octagonal base provides stability. Autoclaving may alter accuracy. Smallest size has clear base; larger sizes have black base.