AlphaTec® 88-394 Natural Rubber Latex Gloves, Unlined, Ansell
Supplier: Ansell Healthcare
120129 120130 120131 120132
32890-365CS 897 CAD
32890-365 32890-366 32890-368 32890-369
AlphaTec® 88-394 Natural Rubber Latex Gloves, Unlined, Ansell
Gloves
Gloves provide excellent dexterity, tensile strength and elasticity, along with chemical, snag, puncture and abrasion resistance.
  • Excellent dexterity and tensile strength
  • Strong chemical and puncture resistance
  • Ideal glove for safety and dexterity in the food processing industry
Use in the agriculture, viticulture, food processing and food service industries. Uses include canning and food processing, food packing and handling, fruit and vegetable processing, light-duty maintenance and clean-up, and general industry.
Certifications: Complies withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Latex/Latex-free Latex
Finish Fishscale
Cuff style Pinked
EN norm 2110