WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Inserts and µLMats, Assembled, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
07-1220MB-101 07-1220MG-101 07-1220MN-101 07-1220MR-101 07-1221MB-101 07-1221MG-101 07-1221MN-101 07-1221MR-101 07-1245MB-101 07-1245MG-101 07-1245MN-101 07-1245MR-101 07-1275MN-101 07-1275MR-101 07-2020MB-201 07-2020MG-201 07-2020MN-201 07-2020MR-201 07-2021MB-201 07-2021MG-201 07-2021MN-201 07-2021MR-201 07-2045MB-201 07-2045MG-201 07-2045MN-201 07-2045MR-201 07-2075MB-201 07-2075MG-201 07-2075MN-201 07-2075MR-201 07-3020MB-300 07-3020MG-300 07-3020MN-300 07-3020MR-300 07-3021MB-300 07-3021MG-300 07-3021MN-300 07-3021MR-300 07-3045MB-300 07-3045MG-300 07-3045MN-300 07-3045MR-300 07-3075MB-300 07-3075MG-300 07-3075MN-300 07-3075MR-300 07-8020MB-1200 07-8020MB-2200 07-8020MG-1200 07-8020MG-2200 07-8020MN-1200 07-8020MN-2200 07-8020MR-1200 07-8020MR-2200 07-8021MB-1200 07-8021MB-2200 07-8021MG-1200 07-8021MG-2200 07-8021MN-1200 07-8021MN-2200 07-8021MR-1200 07-8021MR-2200 07-8045MB-1200 07-8045MB-2200 07-8045MG-1000 07-8045MG-1200 07-8045MG-2200 07-8045MN-1200 07-8045MN-2200 07-8045MR-1200 07-8045MR-2200 07-8075MB-1200 07-8075MB-2200 07-8075MG-1200 07-8075MG-2200 07-8075MN-1200 07-8075MN-2200 07-8075MR-1200 07-8075MR-2200 07-1270MB-101 07-1270MN-101 07-1270MR-101 07-2070MB-201 07-2070MG-201 07-2070MN-201 07-2070MR-201 07-3070MB-300 07-3070MG-300 07-3070MN-300 07-3070MR-300 07-8070MB-1200 07-8070MB-2200 07-8070MG-2200 07-8070MN-1200 07-8070MN-2200 07-8070MR-1200 07-8070MR-2200 07-3070MB-350 07-3070MG-350 07-3070MN-350 07-3070MR-350 07-3075MB-350 07-3075MG-350 07-3075MN-350 07-3075MR-350 07-2020MB-250 07-2020MG-250 07-2020MN-250 07-2020MR-250 07-2021MB-250 07-2021MG-250 07-2021MN-250 07-2021MR-250 07-2045MB-250 07-2045MG-250 07-2045MN-250 07-2045MR-250 07-2070MB-250 07-2070MG-250 07-2070MN-250 07-2070MR-250 07-2075MB-250 07-2075MG-250 07-2075MN-250 07-2075MR-250 07-3020MB-350 07-3020MG-350 07-3020MN-350 07-3020MR-350 07-3021MB-350 07-3021MG-350 07-3021MN-350 07-3021MR-350 07-3045MB-350 07-3045MG-350 07-3045MN-350 07-3045MR-350
97044-962EA 249.28 CAD
97044-962 97044-964 97044-966 97044-968 97044-986 97044-988 97044-990 97044-992 97045-010 97045-012 97045-014 97045-016 97045-040 97045-042 97045-044 97045-046 97045-076 97045-078 97045-080 97045-082 97045-100 97045-102 97045-104 97045-106 97045-124 97045-128 97045-130 97045-132 97045-156 97045-158 97045-160 97045-162 97045-206 97045-208 97045-210 97045-212 97045-230 97045-232 97045-234 97045-236 97045-254 97045-258 97045-260 97045-262 97045-286 97045-288 97045-290 97045-292 97045-424 97045-428 97045-430 97045-432 97045-434 97045-438 97045-440 97045-442 97045-444 97045-448 97045-450 97045-452 97045-454 97045-458 97045-460 97045-462 97045-544 97045-548 97045-550 97045-552 97045-554 97045-558 97045-560 97045-562 97045-564 97045-568 97045-570 97045-572 97045-574 97045-578 97045-580 97045-582 97045-664 97045-668 97045-670 97045-672 97045-674 97045-678 97045-680 97045-682 97045-684 97045-688 97045-690 97045-692 97045-694 97045-698 97045-700 97045-702 97045-788 97045-792 97045-794 97045-796 97045-798 97045-802 97045-804 97045-806 97045-808 97045-812 97045-814 97045-816 97045-818 97045-822 97045-824 97045-826 89231-786 89231-788 89231-790 89231-808 89231-810 89231-812 89231-814 89231-832 89231-834 89231-836 89231-838 89231-888 89231-892 89231-894 89231-896 89231-898 89231-904 89231-906 89231-908 89231-912 89231-914 89231-916 89231-918 89231-922 89231-924 89231-926 30618-408 30618-410 30618-412 30618-414 30618-432 30618-434 30618-436 30618-438 30618-214 30618-216 30618-218 30618-220 30618-238 30618-240 30618-242 30618-244 30618-262 30618-264 30618-266 30618-268 30618-286 30618-288 30618-290 30618-292 30618-310 30618-312 30618-314 30618-316 30618-336 30618-338 30618-340 30618-342 30618-360 30618-362 30618-364 30618-366 30618-384 30618-386 30618-388 30618-390
WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Inserts and µLMats, Assembled, DWK Life Sciences
Microplates Deep Well Microplates
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with glass inserts and color-coded capmats.
  • Variety of volumes available
  • Standard chromatography closures
  • CapMats included
  • Silicone mats are resealable
A comprehensive line of 96-Well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials
Polypropylene μLPlates® have a variety of closures that will stabilize the sample during the analytical sample run and, ifMore Product Information