WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Inserts and Caps, Assembled, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
07-1220MB-101 07-1220G-101-A 07-1220N-101-A 07-1220R-101-A 07-1221B-101-A 07-1221G-101-A 07-1221N-101-A 07-1221R-101-A 07-1245B-101-A 07-1245G-101-A 07-1245N-101-A 07-1245R-101-A 07-1275B-101-A 07-1275G-101-A 07-1275N-101-A 07-1275R-101-A 07-2020B-201-A 07-2020G-201-A 07-2020N-201-A 07-2020R-201-A 07-2021B-201-A 07-2021G-201-A 07-2021N-201-A 07-2021R-201-A 07-2045B-201-A 07-2045G-201-A 07-2045N-201-A 07-2045R-201-A 07-2075B-201-A 07-2075G-201-A 07-2075N-201-A 07-2075R-201-A 07-3020B-300-A 07-3020G-300-A 07-3020N-300-A 07-3020R-300-A 07-3021B-300-A 07-3021G-300-A 07-3021N-300-A 07-3021R-300-A 07-3045B-300-A 07-3045G-300-A 07-3045N-300-A 07-3045R-300-A 07-3075B-300-A 07-3075G-300-A 07-3075N-300-A 07-3075R-300-A 07-8020B-1202-A 07-8020G-1200-A 07-8075B-1200-A 07-8075B-2200-A 07-8075G-1200-A 07-8075G-2200-A 07-8075N-1200-A 07-8075N-2200-A 07-8075R-1200-A 07-8075R-2200-A 07-1270B-101-A 07-1270G-101-A 07-1270N-101-A 07-1270R-101-A 07-2070B-201-A 07-2070G-201-A 07-2070N-201-A 07-2070R-201-A 07-3070B-300-A 07-3070G-300-A 07-3070N-300-A 07-3070R-300-A 07-8070B-1200-A 07-8070B-2200-A 07-8070G-1200-A 07-8070G-2200-A 07-8070N-1200-A 07-8070N-2200-A 07-8070R-1200-A 07-8070R-2200-A 07-3070B-350-A 07-3070G-350-A 07-3070N-350-A 07-3070R-350-A 07-3075B-350-A 07-3075G-350-A 07-3075N-350-A 07-3075R-350-A 07-2020B-250-A 07-2020G-250-A 07-2020N-250-A 07-2020R-250-A 07-2021B-250-A 07-2021G-250-A 07-2021N-250-A 07-2021R-250-A 07-2045B-250-A 07-2045G-250-A 07-2045N-250-A 07-2045R-250-A 07-2070B-250-A 07-2070G-250-A 07-2070N-250-A 07-2070R-250-A 07-2075B-250-A 07-2075G-250-A 07-2075N-250-A 07-2075R-250-A 07-3020B-350-A 07-3020G-350-A 07-3020N-350-A 07-3020R-350-A 07-3021B-350-A 07-3021G-350-A 07-3021N-350-A 07-3021R-350-A 07-3045B-350-A 07-3045G-350-A 07-3045N-350-A 07-3045R-350-A
97044-956PK 248.84 CAD
97044-956 97044-960 97044-972 97044-976 97044-980 97044-984 97044-996 97045-000 97045-004 97045-008 97045-020 97045-024 97045-034 97045-038 97045-050 97045-054 97045-070 97045-074 97045-086 97045-090 97045-094 97045-098 97045-110 97045-114 97045-118 97045-122 97045-136 97045-140 97045-150 97045-154 97045-166 97045-170 97045-200 97045-204 97045-216 97045-220 97045-224 97045-228 97045-240 97045-244 97045-248 97045-252 97045-266 97045-270 97045-280 97045-284 97045-296 97045-300 97045-386 97045-394 97045-398 97045-402 97045-406 97045-414 97045-418 97045-422 97045-466 97045-474 97045-478 97045-482 97045-486 97045-494 97045-498 97045-502 97045-506 97045-514 97045-518 97045-522 97045-526 97045-534 97045-538 97045-542 97045-586 97045-594 97045-598 97045-602 97045-606 97045-614 97045-618 97045-622 97045-626 97045-634 97045-638 97045-642 97045-646 97045-654 97045-658 97045-662 97045-706 97045-714 97045-718 97045-722 97045-726 97045-734 97045-738 97045-742 97045-750 97045-758 97045-762 97045-766 97045-770 97045-778 97045-782 97045-786 97045-830 97045-838 97045-842 97045-846 97045-850 97045-858 97045-862 97045-866 89231-780 89231-784 89231-794 89231-798 89231-802 89231-806 89231-818 89231-822 89231-826 89231-830 89231-842 89231-846 89231-850 89231-858 89231-862 89231-866 89231-870 89231-878 89231-882 89231-886 89231-930 89231-938 89231-942 89231-946 89231-950 89231-958 89231-962 89231-966 30618-402 30618-406 30618-418 30618-422 30618-426 30618-430 30618-442 30618-446 30618-208 30618-212 30618-224 30618-228 30618-232 30618-236 30618-248 30618-252 30618-256 30618-260 30618-272 30618-276 30618-280 30618-284 30618-296 30618-300 30618-304 30618-308 30618-320 30618-324 30618-330 30618-334 30618-346 30618-350 30618-354 30618-358 30618-370 30618-374 30618-378 30618-382 30618-394 30618-398
WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Inserts and Caps, Assembled, DWK Life Sciences
Microplates Deep Well Microplates
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with glass inserts and color-coded caps.
  • Variety of volumes available
  • Standard chromatography closures
  • Caps with septa
  • Silicone mats are resealable
A comprehensive line of 96-Well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials
Polypropylene μLPlates® have a variety of closures that will stabilize the sample during the analytical sample run and, ifMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable