Hydrogen peroxide solution 50%

Supplier: GFS CHEMICALS INC
Danger

43991
30617-946EA 178.87 CAD
30617-946
Hydrogen peroxide solution 50%
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01
Boiling Pt: 150.2
Melting Pt: -0.41
Density: 1.11
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1 II
Merck Index: 14,04798

Order Now

Specification Test Results

Concentration 45-55%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR