SP Bel-Art Write-On™ Label Tape, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13462-0300 F13463-0600 F13480-0050 F13481-0050 F13482-0050 F13480-0075 F13481-0075 F13482-0075 F13480-0100 F13481-0100 F13482-0100 F13483-0050 F13484-0050 F13485-0050 F13486-0050 F13487-0050 F13488-0050 F13483-0075 F13484-0075 F13485-0075 F13486-0075 F13487-0075 F13488-0075 F13483-0100 F13484-0100 F13485-0100 F13486-0100 F13487-0100 F13488-0100 F13490-0050 F13490-0075 F13490-0100
89005-238PK 44.29 CAD
89005-238 89005-240 30617-006 30617-008 30617-010 30617-012 30617-014 30617-016 30617-018 30617-020 30617-022 30617-024 30617-026 30617-028 30617-030 30617-032 30617-034 30617-036 30617-038 30617-040 30617-042 30617-044 30617-046 30617-048 30617-050 30617-052 30617-054 30617-056 30617-058 30617-060 30617-062 30617-064
SP Bel-Art Write-On™ Label Tape, Bel-Art Products, a part of SP
Tapes
Adheres to any clean surface, including Teflon® fluoropolymer resin-coated materials, and peels off without leaving a sticky residue.
  • Resists moisture and most solvents
  • Withstands autoclaving and gamma irradiation
  • Suited for use in low-temperature applications
Pencil, ball-point pen, or solvent ink marker remain legible under the most demanding laboratory conditions
The tape will withstand a maximum temperature of 157°C (315°F) for 30 minutes or 80°C (176°F) for 24 hours. It can be used in temperatures as low as –80°C (–112°F). Tape will withstandMore Product Information